Solid phase extraction cartridges, Sep-Pak® Plus
About this item
These cartridges can used with liquid and gas samples and have the unique capability of reverse flow direction, a useful feature for trace enrichment application.
- Precision design suits to robotic application also more convenient in manual use
- Sorbents are in short and long body configurations
Can be connected to different devices like filter units, positive flow pumps and syringes, vacuum manifolds, automated sample processor, and in-line reaction systems (PET applications).
3 Options Available Below
Product Details & Documents
These cartridges can used with liquid and gas samples and have the unique capability of reverse flow direction, a useful feature for trace enrichment application.
- Precision design suits to robotic application also more convenient in manual use
- Sorbents are in short and long body configurations
Can be connected to different devices like filter units, positive flow pumps and syringes, vacuum manifolds, automated sample processor, and in-line reaction systems (PET applications).
Product Family Options
Product Information
- Column FormatPore sizeParticle SizeCapacityBed weightAvailabilityPrice
- WATEWAT020515Sep-Pak® Plus Short, C18Sep-Pak® Plus Short, C18Cartridge125 Å55 - 105 µm0,7 ml360 mgLog in for availability1 * 50 ST411,00 €Specifications:Marketsource
More
Specifications
Cat. No.WATEWAT020515Marketsource - WATEWAT020520Sep-Pak® Plus Long, SilicaSep-Pak® Plus Long, SilicaCartridge125 Å55 - 105 µm1,6 ml690 mgLog in for availability1 * 50 ST411,00 €Specifications:Marketsource
More
Specifications
Cat. No.WATEWAT020520Marketsource - WATEWAT036810Sep-Pak® Plus Short, tC18Sep-Pak® Plus Short, tC18Cartridge125 Å37 - 55 µm0,8 ml400 mgLog in for availability1 * 50 ST411,00 €Specifications:Marketsource
More
Specifications
Cat. No.WATEWAT036810Marketsource