Petri dish filling system, PetriSwiss PS20 mini
Supplier: BioTool
Certificates
About this item
PetriSwiss PS 20 fills 20 dishes directly in the PetriRack. There is no need for manual stacking or any other manual operations. It is ideal for production of small, special-purpose agar batches. The dishes are safely stacked in the PetriRack and can even be transported, chilled or incubated in the rack. The rack can also be fully autoclaved. The dishes can be taken continuously from the rack. All program data, parameters and operating states are displayed clearly and understandably on a graphic display. The unit is very operator-friendly and is operated using a state-of-the-art touch screen.
- Graphic display with backlighting for perfect read-off
- Easily understandable, user-friendly operation with touch screen
- Operator prompting in German or English, freely selectable by the user
- Fully automatic filling and stacking of the Petri dishes
- High, trouble-free Petri dish throughput
- Dosing quantities, speed, pause time and AntiDrop function programmable
- Stack function and automatic self-test initialisation and run
- Self-monitoring of all device functions
- Easy care and servicing of the individual device components
Specifications
- Catalog No:
- 710-1672
- 710-1676
- 710-1678
- 710-1674
- Model:
- PS20 mini
- PS20 mini
- PS20 mini
- PS20 mini
- Capacity:
- 1-20 dishes per stack
- 1-20 dishes per stack
- 1-20 dishes per stack
- 1-20 dishes per stack
- Capacity (samples/run):
- 20 dishes in 1,5 min
- 20 dishes in 1,5 min
- 20 dishes in 1,5 min
- 20 dishes in 1,5 min
- Volume:
- 15 ml
- 15 ml
- 15 ml
- 15 ml
- Weight:
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- W×D×H:
- 505×305×615 mm
- 505×305×615 mm
- 505×305×615 mm
- 505×305×615 mm