Eco caps
Supplier: VICI JOUR RESEARCH
Certificates
About this item
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
- Compatible with all standard GL 45 thread bottles
- Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
Delivery: The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.