ContactUsLinkComponent
Prepacked size exclusion columns, Micro Bio-Spin™ P-6
Prepacked size exclusion columns, Micro Bio-Spin™ P-6
Catalog # BRAD732-6222
Supplier:  BIO RAD
Prepacked size exclusion columns, Micro Bio-Spin™ P-6
Catalog # BRAD732-6222
Supplier:  BIO RAD
Specifications
  • Description:
    Micro Bio-Spin™ P-6 gel columns, prepacked spin cloumns, with Bio-Gel® P-6 in Tris buffer, 200 collection tubes, with sample volume 10–75 μl and 6,000 MW limit
  • Pk:
    100