ContactUsLinkComponent
Cut resistant gloves, Sharpflex Liner
Cut resistant gloves, Sharpflex Liner
Catalog # DALL2232522/9
Supplier:  HONEYWELL SAFETY
undefined
Cut resistant gloves, Sharpflex Liner
Catalog # DALL2232522/9
Supplier:  HONEYWELL SAFETY
Specifications
  • Glove size:
    9
  • Ambidextrous / hand-specific:
    Ambidextrous
  • Disposable / reusable:
    Reusable
  • Colour:
    Yellow/green
  • Length:
    250 mm
  • Seq:
    03
  • Model:
    Liner
  • EN norm:
    EN 388:2016 (2X4XC), EN 407 (X1XXXX)
  • Cuff style:
    Knit wrist
  • Gauge:
    13
  • Pk:
    10 PAIR
Specifications
About this item
These gloves are knitted from composite and Kevlar® yarn.
  • Knitting technique provides the glove with excellent fit and high dexterity
  • Kevlar® yarn protects against contact heat and cuts
  • Plated polyamide inside: Limit the contact of the skin with composite fibre
  • Ambidextrous
Ideal for handling heavy parts with high cut risks in dry environments.
Certifications: EN 388:2016 (2X4XC), EN 407 (X1XXXX); Cat. II