ContactUsLinkComponent
Blades for safety knives, No. 37040
Blades for safety knives, No. 37040
Catalog # 233-2240
Supplier:  MARTOR ARGENTAX
undefined
Blades for safety knives, No. 37040
Catalog # 233-2240
Supplier:  MARTOR ARGENTAX
Specifications
  • Description:
    Replacement industrial cutter blade no. 37040, square ends, deep-edged
  • L×W:
    43,0×22,2 mm
  • Material:
    Carbon steel
  • Thickness:
    0,40 mm
  • Pk:
    10
Specifications
About this item
Industrial blade for use with safety knives.
  • Deep-edged, two-sided ground-edge
  • Double bevel edge-ground
  • Four-times usable blade
Mountable in:
SECUMAX OPTICUT
SECUMAX RUCK-ZUCK
SECUMAX RUCK-ZUCK SOS
SECUMAX SALVEX SOS
SECUMAX COMBI
SECUMAX NOVEX