LC vials and closures
Supplier: Agilent
Certificates
About this item
Caps and septa
Product Information
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
HPLC columns, Chromolith®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Sleeves for Standard Ground Joints
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
Iron(II) sulphate heptahydrate 99+% ACS
Supplier: Thermo Fisher Scientific
Septa for 16 mm screw top vials
Supplier: Agilent
Supelco® Ascentis® Express C18 HPLC and UHPLC Columns
Supplier: Merck