Orders LinkContactUsLinkComponent
Casserole, deep
Supplier:   CONTACTO
Casserole, deep
Supplier:  CONTACTO
Certificates
Casserole, deep
3 Options Available Below
Product Details & Documents
Stainless steel 18/10.
  • Mirror polished
  • With cool grip handles and pouring rim
  • Sandwich base: 8 mm
Suitable for any cooking method including induction, extra heavy gauge.
Product Family Options
Product Information
  • Capacity
    Ø int.
    Height
    Availability
    Price
  • 2,0 l
    160 mm
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,60 €
    46,60 €
    1 * 1 ST
       46,60 €
  • 4,0 l
    200 mm
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 6,5 l
    240 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €