ContactUsLinkComponent
Cut resistant gloves, K-TEX® 930
Cut resistant gloves, K-TEX® 930
Supplier:  KCL by Honeywell
Certificates
undefined
Cut resistant gloves, K-TEX® 930
Supplier:  KCL by Honeywell
Specifications
  • Catalog No:
  • 112-8300
  • KCHL930.8
  • 112-2992
  • KCHL930.7
  • Model:
  • 930
  • 930
  • 930
  • 930
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Finish:
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388 (3444)
  • EN 388 (3444)
  • EN 388 (3444)
  • EN 388 (3444)
Specifications