Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Clamps
Tubing and Sanitary Clamps
Tubing clamps
Tubing clamps
Tubing clamps
Supplier:
DELTALAB
Certificates
Expand image
Tubing clamps
Supplier:
DELTALAB
Certificates
About this item
Tubing clamps made of acetate.
Corrosion free
Control flow from full open to complete closure
Related Information
Documentation
Certificates
Search by Lot Number
Documentation