Printer for glassware washers
Catalog # 141-0832
Supplier: SMEG
Specifications
- Description:External 24-character dot matrix thermal printer with real-time clock function
- Pk:1
- For:GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW3060SC,
GW4060C, GW4060S, GW4060SC, GW4090, GW4090C