Orders LinkContactUsLinkComponent
Hydrometers
Hydrometers
Hydrometers
Certificates
2 Options Available Below
Product Details & Documents
Both hydrometers (aerometers) are designed for the determination of density by the Archimede's principle.
Certifications: DIN 12791
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • NEUB3910-08
    Hydrometer
    Hydrometer
    0,800 − 0,900 g/ml
    0,002 g/ml
    245 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,80 €
    66,80 €
    1 * 1 ST
       66,80 €
  • NEUB3910-09
    Hydrometer
    Hydrometer
    0,900 − 1,000 g/ml
    0,002  g/ml
    245 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,80 €
    66,80 €
    1 * 1 ST
       66,80 €