Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubes
Test Tubes
Test tubes, with rim
Test tubes, with rim
Test tubes, with rim
Supplier:
NEUBERT VOLUME GLASSWAERE
Certificates
Expand image
Test tubes, with rim
Supplier:
NEUBERT VOLUME GLASSWAERE
About this item
Quartz, clear.
Excellent chemical resistance
Excellent temperature resistance