Bandages, Aluderm®
Supplier: SOEHNGEN
Certificates
About this item
Aluminised wound dressing made of polyester non woven fabric. The final layer is a hydrophobic PP non woven fabric. These bandages feature a no-touch winding.
- Absorbent layer has an absorption capacity of 1400 g/m²
- Will not stick to any wound, breathable
- Bandages do not release fibres
- Strongly absorbent, keeping even the highly secreting wounds dry
- Promotes skin formation and healing, physiologically safe, free of chemical binding materials