Cut resistant gloves, HyFlex® 70-205
Supplier: Ansell
Certificates
About this item
These knitted gloves are designed to provide superior mechanical strength in light to medium-duty handling and production applications. The Kevlar® reinforcement provides a very high cut resistance, offers enhanced comfort and dexterity.
- Neptune® coating: glove fibres repel liquids, avoiding damage to the Kevlar® strands and extending glove life
- Reinforced thumb welt increases abrasion resistance
- Fully washable and can be reused many times
Ideal for applications involving metal and glass.
Certifications: EN 388:2016 (134XB), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- 112-3552
- 112-3554
- 112-3555
- 112-3553
- Model:
- 70-205
- 70-205
- 70-205
- 70-205
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Finish:
- Neptune® coating
- Neptune® coating
- Neptune® coating
- Neptune® coating
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (134XB), EN 407 (X1XXXX)
- EN 388:2016 (134XB), EN 407 (X1XXXX)
- EN 388:2016 (134XB), EN 407 (X1XXXX)
- EN 388:2016 (134XB), EN 407 (X1XXXX)