Order Entry
Germany
ContactUsLinkComponent
Sacrum and coccyx
Sacrum and coccyx
Catalog # BINH1000139
Supplier:  3B Scientific
undefined
Sacrum and coccyx
Catalog # BINH1000139
Supplier:  3B Scientific

Specifications

  • Description:
    Sacrum and coccyx
  • Pk:
    1

Specifications

About this item

Study the anatomy of the human skeleton in detail with 3B skeletal models! This high quality model details the human sacrum and coccyx.

  • Completely assembled

Dimensions: 20×15×15 cm

Weight: 0,2 kg