Heat resistant gloves, PBI®
: JUTEC
These mittens made of PBI® fabric provide protection against contact heat up to 800 °C.
- Cut and abrasion resistant
- Special insulation
- Universal size fits all
Ideal for use in foundries and in the metal industry.
: EN 388, EN 407; Cat. III
- Catalog No:
- JUTEH111B140-PBI
- JUTEH111B130-PBI
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- EN norm:
- EN 388, EN 407
- EN 388, EN 407