Order Entry
Germany
ContactUsLinkComponent
Cleaning wipe, microfiber
Cleaning wipe, microfiber
Catalog # 115-2151
undefined
Cleaning wipe, microfiber
Catalog # 115-2151

Specifications

  • Colour:
    Blue
  • L×W:
    400×380 mm
  • Pk:
    1

Specifications

About this item

These microfibre wipes are ideal for the smear-free cleaning of glass, chrome, plastics and optical lenses.

  • For cleaning without detergents
  • Can be used either wet or dry
  • Washable up to 90 °C
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904