Cleanroom Waste Receptacles
Supplier: Palbam
Certificates
About this item
Stainless steel 304, electropolished finish.
- No need to come in contact with the bin
- No flaps or other devices are attached, as these just increase contact between user and the waste bin
- Free-standing model is mounted on castors which are hidden from view
These round waste receptacles include an internal nylon holder and lid. The lid has a large opening for easy disposal.