Order Entry
Germany
ContactUsLinkComponent
Dispenser for wipes, WDL-39
Dispenser for wipes, WDL-39
Catalog # SCURWDL-39
Supplier:  S-CURVE
undefined
Dispenser for wipes, WDL-39
Catalog # SCURWDL-39
Supplier:  S-CURVE

Specifications

  • Description:
    Dispenser with lid, clear acrylic, for wipes (228,6×228,6 mm)
  • W×D×H:
    241×241×140 mm
  • Pk:
    1

Specifications

About this item

Compact dispenser made of clear acrylic. With lid for easy access from front or top.

  • Full length hinged lid for durability and contamination control
  • Wall mountable or free-standing with rubber pads on bottom corners
  • Flame treated edges for smoothness
  • Packaged in a poly bag