Pallets and sumps for KTC with filling stand, WSP
These pallets and sumps with filling stand are used for storing acids and alkalis.
- Sump capacity of 1000 L
- With chemical resistant PE-inserts for storing acids and alkalis
These pallets and sumps with filling stand are used for storing acids and alkalis.