Cytology brushes
Supplier: DELTALAB
Certificates
About this item
These cytology brushes are ideal for cervical specimen collection, designed for taking cell samples without damaging them.
- Blue ABS shaft
- Includes a soft tip to provide more comfort for patients
- Sterile model is sterilised by ethylene oxide
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904