Liquid micro syringe, VICI Pressure-Lok® C-160
Catalog # SIALZ170860-1EA
Supplier: Merck
Specifications
- For:GC chromatography
- Type:With fixed needle
- Type of tip:Leaktight plunger tip
- Volume:25 µl
- Pk:1
Specifications
About this item
This liquid micro syringe is a general-purpose GC syringe. Each syringe is hand-calibrated for consistent delivery.
- Barrel made of borosilicate glass
- Shaped rear flange provides secure finger grip
The leak tight PTFE plunger tip eliminates freeze-up or residue build-up and allows use with gases (up to 17,2 bar) as well as liquids.
Syringes with removable needles have a pressure tight, low dead-volume needle connection for a leak tight fit.