Conway diffusion cell, SP Bel-Art
Catalog # BELAF409410000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:60 mm
- Colour:Transparent
- Description:Conway diffusion cell chambers
- Length:10 mm
- Material:PP
- Pk:3
Specifications
About this item
Conway diffusion cells made of smooth white molded PP with natural clear cover.
- Obrink modification of the cell
- Knob on the cover used to raise it
- Circular moat of the cell receives the cover sealing off the outermost chamber
- Steam autoclavable at 121 °C
Used for assays of carbon dioxide, ammonia, or alcohol levels in blood or serum using the diffusion method.