Hand cleansers, Estesol® premium
Catalog # EVON22350
Supplier: Deb-STOKO
Specifications
- Description:Hand cleanser, Estesol® premium, 1000 ml hard bottle for use with Stoko Mat® Alu
- Seq:02
- Pk:9
Specifications
About this item
Liquid skin cleanser for light to medium-duty contamination. Ideal for highly-stressed and sensitive skin.
- Contains skin compatible sugar surfactants and natural moisturisers
- Skin friendly pH
- Subtly scented
Colour: Clear
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904