ContactUsLinkComponent
Dialysis bag clips, SP Bel-Art
Dialysis bag clips, SP Bel-Art
Catalog # 229-0003
undefined
Dialysis bag clips, SP Bel-Art
Catalog # 229-0003
Specifications
  • Colour:
    Translucent
  • Description:
    Dialysis bag holder, for fitting onto the side of an open container, beaker or pitcher up to 1⁄4" thick
  • Material:
    PP
  • Pk:
    6
Specifications
About this item
These strong clips made from PP are ideal for suspending dialysis bags in large beakers or pitchers up to 0,63 cm (¼") thick.
  • Bags slide easily into the slot and then the holder can be clipped onto the container’s edge
  • Smooth PP surface may be marked with identifying sequence numbers
  • Autoclavable