Orders LinkContactUsLinkComponent
pH indicator papers
pH indicator papers
Universal pH indicator paper, 5 m roll, 1 - 14
Catalog # SIMESP2000
Supplier:   Industrial Physics
pH indicator papers
Catalog # SIMESP2000
Supplier:  Industrial Physics
pH indicator papers
pH indicator papers
Universal pH indicator paper, 5 m roll, 1 - 14
Price
47,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Universal pH indicator paper on a roll for acid and base (alkaline) test.
  • Fast response time within a minute
  • Easy to read, accurate colour charts
  • High contrast for easy colour chart comparisons
Documents
Specifications
  • pH range:
    1 - 14
  • Description:
    Universal pH indicator paper, 5 m roll