ContactUsLinkComponent
Alcohol meter, Alcotest® 7510
Alcohol meter, Alcotest® 7510
Catalog # DRAE8319760
Supplier:  Dräger
undefined
Alcohol meter, Alcotest® 7510
Catalog # DRAE8319760
Supplier:  Dräger
Specifications
  • Model:
    Alcotest® 7510
  • Range:
    0 - 3,0 mg/L
  • Description:
    Robust handheld alcohol meter
  • Pk:
    1
Specifications
About this item
The Alcotest 7510 is a compact and robust handheld breath alcohol measuring instrument, especially designed for advanced screening applications.
  • Hygienic and easy to use
  • Providing fast, accurate, and reliable results
  • Meets different international regulations and guidelines
Suitable for use by police as well as in commerce and industry as an alcohol screening device, it can also be used for evidential testing purposes, subject to relevant local and market sector approvals.