Bottles with pump vaporiser
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Certificates
About this item
Amber soda glass bottles with pump vaporiser and suitable for chromatography.
- Pump volume 0,12 ml per stroke
- Good chemical resistance
- Amber coloured for light sensitive substances