tert-Butyl methyl malonate 95%
Catalog # H59540.AB
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
Specifications
About this item
:
Boiling Pt:80 °C (11 mmHg)
Density:1.030
Flash Pt:79 °C (174 °F)
:174.19 g/mol
Storage Temperature:Ambient
EINECS:255-919-1
CAS Number:42726-73-8
Formula:C₈H₁₄O₄
MDL Number:MFCD00042856