Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-chlorophenol
3-Bromo-5-chlorophenol
3-Bromo-5-chlorophenol
COBBOT-0189-25G
:
COMBI-BLOCKS
:
3-Bromo-5-chlorophenol
COBBOT-0189-25G
:
COMBI-BLOCKS
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
207.45 g/mol
:
56962-04-0
:
C₆H₄BrClO
:
MFCD07780676