Order Entry
Germany
ContactUsLinkComponent
Trifluoroacetic acid
Trifluoroacetic acid
Catalog # PIER85183
CAS Number:  
undefined
Trifluoroacetic acid
Catalog # PIER85183
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
 :GHS05
 :GHS07
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169