Order Entry
Germany
Orders LinkContactUsLinkComponent
 

5'-Bromo-2'-hydroxyacetophenone ≥98%

5'-Bromo-2'-hydroxyacetophenone ≥98%
Certificates
5'-Bromo-2'-hydroxyacetophenone ≥98%
5'-Bromo-2'-hydroxyacetophenone ≥98%

2 Options Available Below

Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
  • CAS Number:1450-75-5
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00191850
  • Molecular Weight:215.05
  • Melting Pt:56…60 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 46,90 €
    46,90 €
    1 * 10 g
       46,90 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 224,00 €
    224,00 €
    1 * 50 g
       224,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.