

Ninhydrin solution R1 Reag. Ph. Eur.
Artikelnr. REGE1058304
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:178.14 g/mol
Storage Temperature:Ambient
EINECS:207-618-1
UN:2924
ADR:3,II
:GHS02
:GHS05
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791