Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nonyl β-D-glucopyranoside
Nonyl-β-D-glucopyranoside ≥98%
Nonyl-β-D-glucopyranoside ≥98%
Nonyl-β-D-glucopyranoside ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Nonyl-β-D-glucopyranoside ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
69984-73-2
:
Formula:
C₁₅H₃₀O₆
MDL Number:
MFCD00063300
:
306.4 g/mol
Storage Temperature:
Freezer
Related Information
Documentation
Videos
Thermo Scientific Acroseal 25 Years
Documentation