Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ortho-Boric acid
Orthoboric acid 20 g/l in aqueous solution for nitrogen analysis
Orthoboric acid 20 g/l in aqueous solution for nitrogen analysis
Orthoboric acid 20 g/l in aqueous solution for nitrogen analysis
Supplier:
Bernd Kraft
Certificates
Expand image
Orthoboric acid 20 g/l in aqueous solution for nitrogen analysis
Supplier:
Bernd Kraft
About this item
CAS Number:
10043-35-3
:
Formula:
B(OH)₃
MDL Number:
MFCD00011337
:
61.83 g/mol
Storage Temperature:
Ambient
EINECS:
233-139-2