Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyronin G

Pyronin G

Supplier:   Cayman Chemical
Pyronin G
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
  • CAS Number:92-32-0
  • Formula:C₁₇H₁₉ClN₂O
  • MDL Number:MFCD00011725
  • Molecular Weight:302.8 g/mol
  • Merck Index:12,08190
  • Melting Pt:250…260 °C
  • Storage Temperature:Ambient
  • EINECS:202-147-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 46,00 €
    46,00 €
    1 * 1 g
       46,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 90,00 €
    90,00 €
    1 * 5 g
       90,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 154,00 €
    154,00 €
    1 * 10 g
       154,00 €