7-Chloroisatin
Catalog # COBBOR-1448-5G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:181.58 g/mol
:GHS07
CAS Number:7477-63-6
Formula:C₈H₄ClNO₂
MDL Number:MFCD00022796