L(+)-Leucine for SILAC
Catalog # PIER88428
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-522-0
CAS Number:61-90-5
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617