(R)-(-)-1-Methoxy-2-propanol
Catalog # COBBOR-1566-25G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:90.12 g/mol
Storage Temperature:Ambient
UN:3092
ADR:3,III
CAS Number:4984-22-9
:49840-22-9
Formula:C₄H₁₀O₂
MDL Number:MFCD01632587