2-Formyl-5-methoxypyridine
Catalog # ASTA61667
Supplier: ASTATECH
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.1360 g/mol
CAS Number:22187-96-8
Formula:C₇H₇NO₂
MDL Number:MFCD08234867