Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Chloro-4-fluorophenyl)piperidine
1-(3-Chloro-4-fluorophenyl)piperidine
1-(3-Chloro-4-fluorophenyl)piperidine
1-(3-Chloro-4-fluorophenyl)piperidine
Supplier:
COMBI-BLOCKS
Certificates
Expand image
1-(3-Chloro-4-fluorophenyl)piperidine
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
1033201-89-6
:
Formula:
C₁₁H₁₃ClFN
MDL Number:
MFCD10699654
:
213.68 g/mol