Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
Supplier:
FLUOROCHEM
Certificates
Expand image
1-Bromo-2-(difluoromethyl)benzene
Supplier:
FLUOROCHEM
About this item
CAS Number:
845866-82-2
:
Formula:
C₇H₅BrF₂
MDL Number:
MFCD06657962
:
207.0154 g/mol