

4-(Trimethylsilyl)-3-butyn-1-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2117-12-6
:
Formula:C₇H₁₄OSi
MDL Number:MFCD00190217
:142.27 g/mol
Boiling Pt:97…98 °C (24 mmHg)
Density:0.854
Flash Pt:60 °C (140 °F)
Storage Temperature:Ambient
ADR:3,III
:GHS02
:GHS07