

4-(Trimethylsilyl)-3-butyn-1-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2117-12-6
:
Formula:C₇H₁₄OSi
MDL Number:MFCD00190217
:142.27 g/mol
Boiling Pt:97…98 °C (24 mmHg)
Density:0.854 g/cm³
Flash Pt:60 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4540 to 1.4560 (20°C, 589 nm)
- GC:≥97.5 %