Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Cyano-2-methylaniline
3-Cyano-2-methylaniline
3-Cyano-2-methylaniline
3-Cyano-2-methylaniline
Catalog # ENAMEN300-112929
Supplier:
SIA ENAMINE
CAS Number:
Expand image
3-Cyano-2-methylaniline
Catalog # ENAMEN300-112929
Supplier:
SIA ENAMINE
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
500 mg
Seq:
2725
Specifications
About this item
Product Details
:
CAS Number:
69022-35-1
Formula:
C₈H₈N₂
Product Details