Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-(Trifluoromethyl)phenyl)thiourea
1-(4-(Trifluoromethyl)phenyl)thiourea
1-(4-(Trifluoromethyl)phenyl)thiourea
1-(4-(Trifluoromethyl)phenyl)thiourea
Supplier:
MENAI
Certificates
Expand image
1-(4-(Trifluoromethyl)phenyl)thiourea
Supplier:
MENAI
About this item
CAS Number:
1736-72-7
:
Formula:
C₈H₇F₃N₂S
MDL Number:
MFCD00041189
:
220.22 g/mol