3-(4-Methoxyphenoxy)propionic acid
Glass bottle for solids, 1 g
Catalog # ENAMEN30000079.1G
Marketsource
Supplier: SIA ENAMINE
3-(4-Methoxyphenoxy)propionic acid
Catalog # ENAMEN30000079.1G
Supplier: SIA ENAMINE
Marketsource
CAS Number: 20811-60-3
Jump to:
Product Details & Documents
Synonyms: 3-(4-Methoxyphenoxy)propanoic acid
- CAS Number:20811-60-3
- Formula:C₁₀H₁₂O₄
- MDL Number:MFCD00020515
- Molecular Weight:196.2 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



