(Chloromethyl)dimethylethylsilane
Catalog # SILNSILS04025.1
Supplier: CHEMPUR SILANE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:3121-77-5
Formula:C₃H₉ClSi
MDL Number:MFCD00040861