Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-2-methoxypyridine
5-Iodo-2-methoxypyridine ≥98%
5-Iodo-2-methoxypyridine ≥98%
Irritant
5-Iodo-2-methoxypyridine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5-Iodo-2-methoxypyridine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
13472-61-2
:
Formula:
C₆H₆INO
MDL Number:
MFCD07781180
:
235.03 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH50135
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation